Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nadsyn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ305343.1 | - | 7 - 8 | 647 | 70 | |
AK020264.1 | - | 7 - 8 | 647 | 70 | ||
AK041101.1 | - | 7 - 8 | 634 | 70 | ||
AK042389.1 | - | 7 - 8 | 607 | 70 | ||
AK045774.1 | - | 7 - 8 | 634 | 70 | ||
AK046491.1 | - | 7 - 8 | 634 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |