Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tada1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030245.3 | NP_084521.1 | 6 - 7 | 930 | 60 | |
GenBank mRNA | AK013599.1 | - | 6 - 7 | 697 | 60 | |
AK151132.1 | - | 6 - 7 | 792 | 60 | ||
AK180851.1 | - | 1 - 2 | 62 | 60 | ||
AK206969.1 | - | 62 | 60 | |||
BC002307.1 | - | 6 - 7 | 732 | 3592046 | 60 | |
BC027337.1 | - | 6 - 7 | 797 | 3995355 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |