Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133981.2 | NP_598742.1 | 12 - 13 | 1612 | 68 | |
NR_045601.1 | 11 - 12 | 1541 | 68 | |||
GenBank mRNA | AK050335.1 | - | 12 - 13 | 1533 | 68 | |
AK054293.1 | - | 12 - 13 | 1518 | 68 | ||
AK086674.1 | - | 12 - 13 | 1552 | 68 | ||
AK149592.1 | - | 11 - 12 | 1463 | 68 | ||
AK189486.1 | - | 1 - 2 | 58 | 68 | ||
BC021791.1 | - | 12 - 13 | 1507 | 4459738 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |