Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134059.2 | NP_598820.2 | 3 - 4 | 448 | 94 | |
GenBank mRNA | AK151427.1 | - | 3 - 4 | 322 | 94 | |
AK156957.1 | - | 3 - 4 | 316 | 94 | ||
AK168043.1 | - | 3 - 4 | 321 | 94 | ||
BC011308.1 | - | 3 - 4 | 302 | 3256762 | 94 | |
BC023711.1 | - | 3 - 4 | 322 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |