Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bre | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144541.1 | NP_653124.1 | 9 - 10 | 904 | 65 | |
NM_181279.1 | NP_851796.1 | 11 - 12 | 1015 | 65 | ||
NM_181280.1 | NP_851797.1 | 10 - 11 | 913 | 65 | ||
NM_181281.1 | NP_851798.1 | 8 - 9 | 748 | 65 | ||
NM_181282.1 | NP_851799.1 | 9 - 10 | 825 | 65 | ||
GenBank mRNA | AF440752.1 | - | 9 - 10 | 904 | 65 | |
AF527952.1 | - | 11 - 12 | 1015 | 65 | ||
AF527953.1 | - | 9 - 10 | 825 | 65 | ||
AF527954.1 | - | 9 - 10 | 835 | 65 | ||
AF527955.1 | - | 8 - 9 | 748 | 65 | ||
AF538925.1 | - | 10 - 11 | 913 | 65 | ||
AK080991.1 | - | 9 - 10 | 940 | 65 | ||
AK156929.1 | - | 9 - 10 | 996 | 65 | ||
BC061000.1 | - | 9 - 10 | 980 | 30305062 | 65 | |
BC100565.1 | - | 5 - 6 | 459 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |