Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crtac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145123.4 | NP_660105.3 | 6 - 7 | 1004 | 67 | |
GenBank mRNA | AJ421516.1 | - | 6 - 7 | 854 | 67 | |
AK032328.1 | - | 6 - 7 | 869 | 67 | ||
AK049801.1 | - | 6 - 7 | 975 | 67 | ||
AK083221.1 | - | 6 - 7 | 1008 | 67 | ||
AK149020.1 | - | 6 - 7 | 1010 | 67 | ||
AK149069.1 | - | 6 - 7 | 1005 | 67 | ||
BC024472.1 | - | 3 - 4 | 440 | 4976509 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |