Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170556.1 | NP_001164027.1 | 1 - 2 | 187 | 78 | |
NM_145401.2 | NP_663376.2 | 5 - 6 | 1269 | 78 | ||
GenBank mRNA | AK010703.1 | - | 1 - 2 | 129 | 78 | |
AK154296.1 | - | 5 - 6 | 1270 | 78 | ||
AK165834.1 | - | 1 - 2 | 129 | 78 | ||
AY348864.1 | - | 1 - 2 | 120 | 78 | ||
BC015283.1 | - | 5 - 6 | 1057 | 4238045 | 78 | |
BQ572978.1 | - | 1 - 2 | 120 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus 1700 Probe spans exons |