Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163505.1 | NP_001156977.1 | 7 - 8 | 984 | 109 | |
NM_146091.4 | NP_666203.3 | 7 - 8 | 886 | 109 | ||
GenBank mRNA | AK028842.1 | - | 7 - 8 | 747 | 109 | |
AK030660.1 | - | 7 - 8 | 746 | 109 | ||
AK077752.1 | - | 7 - 8 | 749 | 109 | ||
AK147720.1 | - | 7 - 8 | 844 | 109 | ||
AK170550.1 | - | 7 - 8 | 790 | 109 | ||
BC002149.1 | - | 4 - 5 | 276 | 109 | ||
BC017138.1 | - | 7 - 8 | 735 | 4485115 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |