Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700009152.mm.1 |
TC0700000997.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700009152.mm.1 |
NM_001191001.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009152.mm.1 |
NM_001191001.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009152.mm.1 |
NM_152815.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009152.mm.1 |
NM_152815.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009152.mm.1 |
NR_034030.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009152.mm.1 |
NR_034030.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009151.mm.1 |
TC0700000997.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700009151.mm.1 |
NM_001191001.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009151.mm.1 |
NM_001191001.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009151.mm.1 |
NM_152815.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009151.mm.1 |
NM_152815.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009151.mm.1 |
NR_034030.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009151.mm.1 |
NR_034030.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009158.mm.1 |
TC0700000997.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700009158.mm.1 |
NM_001191001.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009158.mm.1 |
NM_001191001.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009158.mm.1 |
NM_152815.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009158.mm.1 |
NM_152815.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009150.mm.1 |
TC0700000997.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700009150.mm.1 |
NM_001191001.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009150.mm.1 |
NM_001191001.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009150.mm.1 |
NM_152815.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009150.mm.1 |
NM_152815.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009150.mm.1 |
NR_034030.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009150.mm.1 |
NR_034030.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009154.mm.1 |
TC0700000997.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700009154.mm.1 |
NM_001191001.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009154.mm.1 |
NM_001191001.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009154.mm.1 |
NM_152815.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009154.mm.1 |
NM_152815.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009154.mm.1 |
NR_034030.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700009154.mm.1 |
NR_034030.1 |
Refseq |