Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007798.3 | NP_031824.1 | 7 - 8 | 804 | 81 | |
GenBank mRNA | AK002456.1 | - | 7 - 8 | 753 | 81 | |
AK083393.1 | - | 7 - 8 | 804 | 81 | ||
AK147192.1 | - | 7 - 8 | 751 | 81 | ||
AK149884.1 | - | 7 - 8 | 753 | 81 | ||
AK151790.1 | - | 7 - 8 | 751 | 81 | ||
AK153047.1 | - | 7 - 8 | 750 | 81 | ||
AK159992.1 | - | 7 - 8 | 789 | 81 | ||
AK167361.1 | - | 7 - 8 | 785 | 81 | ||
AK170959.1 | - | 7 - 8 | 777 | 81 | ||
AK191341.1 | - | 3 - 4 | 269 | 81 | ||
AK191739.1 | - | 3 - 4 | 290 | 81 | ||
AK191997.1 | - | 3 - 4 | 258 | 81 | ||
AK192159.1 | - | 3 - 4 | 286 | 81 | ||
AK192561.1 | - | 3 - 4 | 286 | 81 | ||
AK194693.1 | - | 3 - 4 | 252 | 81 | ||
AK195254.1 | - | 3 - 4 | 251 | 81 | ||
AK196531.1 | - | 3 - 4 | 255 | 81 | ||
AK197104.1 | - | 3 - 4 | 286 | 81 | ||
AK197767.1 | - | 3 - 4 | 262 | 81 | ||
AK198000.1 | - | 3 - 4 | 254 | 81 | ||
AK198001.1 | - | 3 - 4 | 286 | 81 | ||
AK198619.1 | - | 3 - 4 | 287 | 81 | ||
AK198709.1 | - | 3 - 4 | 81 | |||
AK199167.1 | - | 2 - 3 | 248 | 81 | ||
AK201305.1 | - | 3 - 4 | 287 | 81 | ||
AK201756.1 | - | 3 - 4 | 286 | 81 | ||
AK202640.1 | - | 3 - 4 | 286 | 81 | ||
AK209352.1 | - | 3 - 4 | 288 | 81 | ||
AK213062.1 | - | 3 - 4 | 286 | 81 | ||
BC006656.1 | - | 7 - 8 | 795 | 3500700 | 81 | |
M14222.1 | - | 6 - 7 | 709 | 81 | ||
S69034.1 | - | 7 - 8 | 800 | 81 | ||
Y18463.1 | - | 3 - 4 | 363 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |