Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007806.3 | NP_031832.2 | 4 - 5 | 359 | 81 | |
GenBank mRNA | AK021200.1 | - | 4 - 5 | 347 | 81 | |
AK078854.1 | - | 4 - 5 | 330 | 81 | ||
AK131580.1 | - | 4 - 5 | 328 | 81 | ||
AK152414.1 | - | 4 - 5 | 350 | 81 | ||
AK171748.1 | - | 4 - 5 | 328 | 81 | ||
BC026791.1 | - | 4 - 5 | 310 | 3980538 | 81 | |
BC060635.1 | - | 3 - 4 | 4365 | 81 | ||
CF738398.1 | - | 4 - 5 | 342 | 81 | ||
EU791539.1 | - | 4 - 5 | 289 | 81 | ||
M31775.1 | - | 4 - 5 | 282 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |