Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eps8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271587.1 | NP_001258516.1 | 15 - 16 | 1883 | 79 | |
NM_001271588.1 | NP_001258517.1 | 15 - 16 | 1814 | 79 | ||
NM_001271589.1 | NP_001258518.1 | 8 - 9 | 884 | 79 | ||
NM_001271595.1 | NP_001258524.1 | 15 - 16 | 1888 | 79 | ||
NM_007945.3 | NP_031971.2 | 16 - 17 | 1889 | 79 | ||
GenBank mRNA | AK136246.1 | - | 6 - 7 | 655 | 79 | |
AK147877.1 | - | 16 - 17 | 1817 | 79 | ||
AK149683.1 | - | 15 - 16 | 1677 | 79 | ||
AK155022.1 | - | 15 - 16 | 1649 | 79 | ||
AK165684.1 | - | 8 - 9 | 887 | 79 | ||
AK166156.1 | - | 16 - 17 | 1724 | 79 | ||
BC016890.1 | - | 15 - 16 | 1900 | 4240899 | 79 | |
L21671.1 | - | 15 - 16 | 1804 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |