Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007946.2 | NP_031972.2 | 1 - 2 | 201 | 101 | |
GenBank mRNA | BC052881.1 | - | 1 - 2 | 183 | 101 | |
D78353.1 | - | 1 - 2 | 207 | 101 | ||
L77979.1 | - | 1 - 2 | 185 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |