Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300014038.mm.1 |
NM_001042673.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014038.mm.1 |
NM_001282942.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014038.mm.1 |
NM_007985.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014038.mm.1 |
TC1300002184.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300014028.mm.1 |
NM_001282942.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014028.mm.1 |
NM_007985.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014028.mm.1 |
TC1300002184.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300014030.mm.1 |
NM_001282942.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014030.mm.1 |
NM_007985.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014030.mm.1 |
TC1300002184.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300014035.mm.1 |
NM_001042673.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014035.mm.1 |
NM_001282942.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014035.mm.1 |
NM_007985.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014035.mm.1 |
TC1300002184.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300014031.mm.1 |
NM_001042673.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014031.mm.1 |
NM_001282942.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014031.mm.1 |
NM_007985.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014031.mm.1 |
TC1300002184.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300014027.mm.1 |
NM_001282942.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014027.mm.1 |
NM_007985.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014027.mm.1 |
TC1300002184.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300014029.mm.1 |
NM_001282942.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014029.mm.1 |
NM_007985.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014029.mm.1 |
TC1300002184.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300014023.mm.1 |
NM_001042673.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014023.mm.1 |
TC1300002184.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300014036.mm.1 |
NM_001042673.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014036.mm.1 |
NM_001282942.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014036.mm.1 |
NM_007985.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014036.mm.1 |
TC1300002184.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300014025.mm.1 |
NM_001282942.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014025.mm.1 |
NM_007985.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300014025.mm.1 |
TC1300002184.mm.1 |
MTA TC |