Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008030.1 | NP_032056.1 | 6 - 7 | 861 | 75 | |
GenBank mRNA | AK149440.1 | - | 6 - 7 | 888 | 75 | |
U87147.1 | - | 6 - 7 | 861 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |