Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H1f0 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008197.3 | NP_032223.2 | 1 | 1026 | 71 | |
GenBank mRNA | AK051213.1 | - | 1 | 789 | 71 | |
AK077473.1 | - | 1 | 703 | 71 | ||
AK090029.1 | - | 1 | 72 | |||
AK145675.1 | - | 1 | 678 | 71 | ||
AK146077.1 | - | 1 | 789 | 71 | ||
AK153990.1 | - | 1 | 790 | 71 | ||
AK155312.1 | - | 1 | 721 | 71 | ||
BC003830.1 | - | 1 | 775 | 3600538 | 71 | |
BC011493.1 | - | 1 | 773 | 4166167 | 71 | |
BC110361.1 | - | 1 | 778 | 5343233 | 71 | |
X13171.1 | - | 1 | 756 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon |