Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il18r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008365.2 | NP_032391.1 | 10 - 11 | 1644 | 106 | |
GenBank mRNA | U43673.1 | - | 10 - 11 | 1644 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mus musculus Inventoried Probe spans exons |