Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009745.2 | NP_033875.2 | 4 - 5 | 575 | 66 | |
GenBank mRNA | AJ011145.1 | - | 4 - 5 | 455 | 66 | |
AK131643.1 | - | 4 - 5 | 471 | 66 | ||
AK155076.1 | - | 4 - 5 | 457 | 66 | ||
BC011471.1 | - | 4 - 5 | 552 | 3660754 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |