Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009849.2 | NP_033979.2 | 5 - 6 | 878 | 87 | |
GenBank mRNA | AF042811.1 | - | 5 - 6 | 802 | 87 | |
AK002553.1 | - | 5 - 6 | 850 | 87 | ||
AK172281.1 | - | 5 - 6 | 826 | 87 | ||
AY376711.1 | - | 5 - 6 | 811 | 87 | ||
BC011241.1 | - | 5 - 6 | 810 | 3582550 | 87 | |
U91511.1 | - | 4 - 5 | 685 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |