Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009908.2 | NP_034038.2 | 6 - 7 | 1034 | 121 | |
GenBank mRNA | AJ006215.1 | - | 6 - 7 | 947 | 121 | |
AK087150.1 | - | 3 - 4 | 336 | 121 | ||
AK155730.1 | - | 6 - 7 | 987 | 121 | ||
BC004606.1 | - | 1 - 2 | 151 | 121 | ||
BC031500.1 | - | 6 - 7 | 963 | 2647633 | 121 | |
BC063776.1 | - | 6 - 7 | 963 | 6398450 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |