Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009984.3 | NP_034114.1 | 6 - 7 | 871 | 68 | |
GenBank mRNA | AF121837.1 | - | 6 - 7 | 835 | 68 | |
AF121838.1 | - | 6 - 7 | 835 | 68 | ||
AF121839.1 | - | 6 - 7 | 835 | 68 | ||
AK002221.1 | - | 6 - 7 | 871 | 68 | ||
AK011589.1 | - | 6 - 7 | 871 | 68 | ||
AK049466.1 | - | 6 - 7 | 872 | 68 | ||
AK136341.1 | - | 5 - 6 | 712 | 68 | ||
AK145051.1 | - | 6 - 7 | 872 | 68 | ||
AK147142.1 | - | 6 - 7 | 873 | 68 | ||
AK150452.1 | - | 6 - 7 | 872 | 68 | ||
AK151169.1 | - | 6 - 7 | 872 | 68 | ||
AK153076.1 | - | 6 - 7 | 872 | 68 | ||
AK153417.1 | - | 6 - 7 | 872 | 68 | ||
AK159511.1 | - | 6 - 7 | 872 | 68 | ||
AK160100.1 | - | 6 - 7 | 872 | 68 | ||
AK161538.1 | - | 6 - 7 | 872 | 68 | ||
AK169394.1 | - | 6 - 7 | 872 | 68 | ||
AK169712.1 | - | 6 - 7 | 872 | 68 | ||
BC068163.1 | - | 6 - 7 | 857 | 30359160 | 68 | |
J02583.1 | - | 6 - 7 | 848 | 68 | ||
X04392.1 | - | 3 - 4 | 525 | 68 | ||
X06086.1 | - | 6 - 7 | 857 | 68 | ||
Y18465.1 | - | 2 - 3 | 387 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |