Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S1pr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010101.4 | NP_034231.1 | 1 - 2 | 329 | 65 | |
GenBank mRNA | AB028143.1 | - | 1 - 2 | 197 | 65 | |
AK028043.1 | - | 1 - 2 | 186 | 65 | ||
AK029852.1 | - | 1 - 2 | 185 | 65 | ||
AK030134.1 | - | 1 - 2 | 184 | 65 | ||
AK047268.1 | - | 1 - 2 | 177 | 65 | ||
AK078443.1 | - | 1 - 2 | 183 | 65 | ||
AK081919.1 | - | 1 - 2 | 174 | 65 | ||
AK083405.1 | - | 1 - 2 | 163 | 65 | ||
AK084944.1 | - | 1 - 2 | 184 | 65 | ||
AK085180.1 | - | 1 - 2 | 184 | 65 | ||
AK087076.1 | - | 1 - 2 | 184 | 65 | ||
AK087139.1 | - | 1 - 2 | 182 | 65 | ||
BC064007.1 | - | 1 - 2 | 169 | 30244496 | 65 | |
BC068176.1 | - | 1 - 2 | 169 | 30613259 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |