Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmo5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161763.1 | NP_001155235.1 | 7 - 8 | 1330 | 91 | |
NM_001161765.1 | NP_001155237.1 | 6 - 7 | 1413 | 91 | ||
NM_010232.4 | NP_034362.2 | 7 - 8 | 1314 | 91 | ||
GenBank mRNA | AK133675.1 | - | 6 - 7 | 1395 | 91 | |
AK137645.1 | - | 3 - 4 | 596 | 91 | ||
AK147040.1 | - | 7 - 8 | 1307 | 91 | ||
BC022991.1 | - | 7 - 8 | 1277 | 5123876 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |