Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100035557.mm.1 |
NM_001077696.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035557.mm.1 |
NM_001077696.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035557.mm.1 |
NM_001284248.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035557.mm.1 |
NM_001284249.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035557.mm.1 |
NM_001284250.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035557.mm.1 |
NM_010412.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035557.mm.1 |
NM_010412.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035557.mm.1 |
TC1100003829.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100035554.mm.1 |
NM_001077696.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035554.mm.1 |
NM_001077696.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035554.mm.1 |
NM_001284248.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035554.mm.1 |
NM_001284249.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035554.mm.1 |
NM_001284250.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035554.mm.1 |
NM_010412.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035554.mm.1 |
NM_010412.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035554.mm.1 |
TC1100003829.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100035553.mm.1 |
NM_001077696.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035553.mm.1 |
NM_001077696.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035553.mm.1 |
NM_001284248.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035553.mm.1 |
NM_001284249.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035553.mm.1 |
NM_001284250.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035553.mm.1 |
NM_010412.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035553.mm.1 |
NM_010412.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035553.mm.1 |
TC1100003829.mm.1 |
MTA TC |