Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elavl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010485.3 | NP_034615.2 | 2 - 3 | 406 | 90 | |
GenBank mRNA | AK034536.1 | - | 2 - 3 | 393 | 90 | |
AK075818.1 | - | 2 - 3 | 368 | 90 | ||
AK078333.1 | - | 2 - 3 | 722 | 90 | ||
AK080365.1 | - | 2 - 3 | 364 | 90 | ||
AK089088.1 | - | 1 - 2 | 113 | 90 | ||
AK148528.1 | - | 2 - 3 | 334 | 90 | ||
AK161667.1 | - | 1 - 2 | 123 | 90 | ||
AK180399.1 | - | 1 - 2 | 147 | 90 | ||
AK180790.1 | - | 1 - 2 | 147 | 90 | ||
AK183909.1 | - | 1 - 2 | 147 | 90 | ||
AK197032.1 | - | 1 - 2 | 147 | 90 | ||
AK203188.1 | - | 1 - 2 | 147 | 90 | ||
U65735.1 | - | 1 - 2 | 175 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |