Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elavl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177883.1 | NP_001171354.1 | 3 - 4 | 902 | 86 | |
NM_010486.2 | NP_034616.1 | 3 - 4 | 497 | 86 | ||
NM_207685.1 | NP_997568.1 | 4 - 5 | 609 | 86 | ||
NM_207686.1 | NP_997569.1 | 3 - 4 | 497 | 86 | ||
GenBank mRNA | AK141919.1 | - | 3 - 4 | 778 | 86 | |
AK149201.1 | - | 3 - 4 | 714 | 86 | ||
AY035378.1 | - | 2 - 3 | 346 | 86 | ||
AY035379.1 | - | 2 - 3 | 346 | 86 | ||
AY035380.1 | - | 2 - 3 | 346 | 86 | ||
BC046598.1 | - | 3 - 4 | 421 | 6312245 | 86 | |
BC049125.1 | - | 4 - 5 | 609 | 6821133 | 86 | |
BC058393.1 | - | 4 - 5 | 609 | 6821133 | 86 | |
U29088.1 | - | 3 - 4 | 496 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |