Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010636.3 | NP_034766.2 | 3 - 4 | 704 | 101 | |
GenBank mRNA | BC067408.1 | - | 3 - 4 | 704 | 30091275 | 101 |
Y14295.1 | - | 3 - 4 | 688 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |