Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmtf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110327.1 | NP_001103797.1 | 1 - 2 | 165 | 59 | |
NM_011806.3 | NP_035936.3 | 1 - 2 | 165 | 59 | ||
GenBank mRNA | AK029147.1 | - | 1 - 2 | 107 | 59 | |
AK030508.1 | - | 1 - 2 | 134 | 59 | ||
AK031887.1 | - | 1 - 2 | 106 | 59 | ||
AK033208.1 | - | 1 - 2 | 131 | 59 | ||
AK039563.1 | - | 1 - 2 | 133 | 59 | ||
AK040107.1 | - | 1 - 2 | 123 | 59 | ||
AK046201.1 | - | 1 - 2 | 61 | 59 | ||
AK078015.1 | - | 1 - 2 | 66 | 59 | ||
AK145803.1 | - | 1 - 2 | 127 | 59 | ||
AK160595.1 | - | 1 - 2 | 138 | 59 | ||
BC010272.1 | - | 1 - 2 | 109 | 59 | ||
BC045141.1 | - | 1 - 2 | 108 | 6307203 | 59 | |
BC060109.1 | - | 1 - 2 | 88 | 59 | ||
BY247901.1 | - | 1 - 2 | 165 | 59 | ||
U70017.1 | - | 1 - 2 | 120 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |