Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ksr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013571.2 | NP_038599.1 | 12 - 13 | 1601 | 81 | |
GenBank mRNA | AK036566.1 | - | 12 - 13 | 1657 | 81 | |
AK137532.1 | - | 11 - 12 | 1338 | 81 | ||
U43585.1 | - | 12 - 13 | 1608 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mus musculus Probe spans exons |