Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbnl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146308.1 | NP_001139780.1 | 2 - 3 | 217 | 84 | |
NM_001146309.1 | NP_001139781.1 | 2 - 3 | 217 | 84 | ||
NM_013810.3 | NP_038838.1 | 2 - 3 | 217 | 84 | ||
GenBank mRNA | AK046073.1 | - | 2 - 3 | 197 | 84 | |
AK053796.1 | - | 2 - 3 | 213 | 84 | ||
AK078082.1 | - | 2 - 3 | 184 | 84 | ||
AK134136.1 | - | 2 - 3 | 184 | 84 | ||
AK134487.1 | - | 2 - 3 | 184 | 84 | ||
AK140752.1 | - | 2 - 3 | 184 | 84 | ||
AK142818.1 | - | 2 - 3 | 167 | 84 | ||
AK146920.1 | - | 2 - 3 | 184 | 84 | ||
AK151096.1 | - | 2 - 3 | 197 | 84 | ||
AK152110.1 | - | 2 - 3 | 182 | 82 | ||
AK152204.1 | - | 2 - 3 | 197 | 84 | ||
AK153072.1 | - | 2 - 3 | 197 | 84 | ||
AK153389.1 | - | 2 - 3 | 171 | 84 | ||
AK159984.1 | - | 2 - 3 | 194 | 84 | ||
AK161973.1 | - | 2 - 3 | 197 | 84 | ||
AK167956.1 | - | 2 - 3 | 157 | 84 | ||
AK168898.1 | - | 2 - 3 | 194 | 84 | ||
AK172471.1 | - | 2 - 3 | 174 | 84 | ||
AK199448.1 | - | 2 - 3 | 94 | 84 | ||
AY098595.1 | - | 2 - 3 | 140 | 84 | ||
BC046430.1 | - | 2 - 3 | 182 | 4986658 | 84 | |
BY293844.1 | - | 2 - 3 | 217 | 84 | ||
U58884.1 | - | 2 - 3 | 170 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |