Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbnl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146308.1 | NP_001139780.1 | 9 - 10 | 937 | 74 | |
NM_001146309.1 | NP_001139781.1 | 9 - 10 | 925 | 74 | ||
NM_013810.3 | NP_038838.1 | 9 - 10 | 928 | 74 | ||
GenBank mRNA | AK046073.1 | - | 9 - 10 | 905 | 74 | |
AK053796.1 | - | 9 - 10 | 921 | 74 | ||
AK078082.1 | - | 9 - 10 | 892 | 74 | ||
AK134136.1 | - | 9 - 10 | 892 | 74 | ||
AK134487.1 | - | 9 - 10 | 904 | 74 | ||
AK140752.1 | - | 9 - 10 | 895 | 74 | ||
AK142818.1 | - | 9 - 10 | 875 | 74 | ||
AK146920.1 | - | 9 - 10 | 892 | 74 | ||
AK151096.1 | - | 9 - 10 | 905 | 74 | ||
AK152110.1 | - | 9 - 10 | 893 | 74 | ||
AK152204.1 | - | 9 - 10 | 905 | 74 | ||
AK153072.1 | - | 9 - 10 | 904 | 74 | ||
AK153389.1 | - | 9 - 10 | 882 | 74 | ||
AK159984.1 | - | 9 - 10 | 902 | 74 | ||
AK161973.1 | - | 8 - 9 | 1199 | 74 | ||
AK167956.1 | - | 9 - 10 | 864 | 74 | ||
AK168898.1 | - | 9 - 10 | 902 | 74 | ||
AK172471.1 | - | 9 - 10 | 882 | 74 | ||
AY098595.1 | - | 9 - 10 | 860 | 74 | ||
BC046430.1 | - | 9 - 10 | 890 | 4986658 | 74 | |
U58884.1 | - | 9 - 10 | 881 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |