Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enpp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136077.2 | NP_001129549.1 | 2 - 3 | 315 | 57 | |
NM_001285994.1 | NP_001272923.1 | 2 - 3 | 315 | 57 | ||
NM_001285995.1 | NP_001272924.1 | 2 - 3 | 315 | 57 | ||
NM_015744.3 | NP_056559.2 | 2 - 3 | 315 | 57 | ||
GenBank mRNA | AF123542.1 | - | 2 - 3 | 171 | 57 | |
AK038940.1 | - | 2 - 3 | 295 | 57 | ||
AK088491.1 | - | 2 - 3 | 188 | 57 | ||
AK161144.1 | - | 2 - 3 | 188 | 57 | ||
AK163881.1 | - | 2 - 3 | 188 | 57 | ||
BC003264.1 | - | 2 - 3 | 192 | 3499038 | 57 | |
BC058759.1 | - | 2 - 3 | 278 | 6491897 | 57 | |
EU131009.1 | - | 2 - 3 | 130 | 57 | ||
EU131010.1 | - | 2 - 3 | 130 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |