Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ercc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015769.2 | NP_056584.2 | 6 - 7 | 1293 | 118 | |
GenBank mRNA | AF189285.1 | - | 6 - 7 | 1293 | 118 | |
AK138588.1 | - | 6 - 7 | 1087 | 118 | ||
AK149326.1 | - | 6 - 7 | 1126 | 118 | ||
BC026792.1 | - | 6 - 7 | 1084 | 4911382 | 118 | |
BC051036.1 | - | 5 - 6 | 824 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |