Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700006678.mm.1 |
NM_001190449.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006678.mm.1 |
NM_001190449.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006678.mm.1 |
NM_016765.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006678.mm.1 |
NM_016765.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006678.mm.1 |
TC1700000663.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700006682.mm.1 |
NM_001190449.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006682.mm.1 |
NM_001190449.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006682.mm.1 |
NM_016765.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006682.mm.1 |
NM_016765.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006682.mm.1 |
TC1700000663.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700006680.mm.1 |
NM_001190449.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006680.mm.1 |
NM_001190449.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006680.mm.1 |
NM_016765.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006680.mm.1 |
NM_016765.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006680.mm.1 |
TC1700000663.mm.1 |
MTA TC |