Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camkk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018883.2 | NP_061371.2 | 4 - 5 | 629 | 86 | |
GenBank mRNA | AF117384.1 | - | 4 - 5 | 584 | 86 | |
AK144483.1 | - | 4 - 5 | 630 | 86 | ||
BC017529.1 | - | 4 - 5 | 594 | 4924656 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |