Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145979.1 | NP_001139451.1 | 10 - 11 | 1538 | 59 | |
NM_019581.3 | NP_062527.2 | 10 - 11 | 1573 | 59 | ||
GenBank mRNA | AB024573.1 | - | 8 - 9 | 1206 | 59 | |
AF168991.1 | - | 10 - 11 | 1478 | 59 | ||
AK087511.1 | - | 10 - 11 | 1444 | 59 | ||
AK146415.1 | - | 10 - 11 | 1523 | 59 | ||
BC049089.1 | - | 10 - 11 | 1534 | 5720710 | 59 | |
BC051958.1 | - | 10 - 11 | 1534 | 5720710 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |