Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gltp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019821.2 | NP_062795.2 | 2 - 3 | 239 | 63 | |
GenBank mRNA | AF209703.1 | - | 2 - 3 | 165 | 63 | |
AK157313.1 | - | 2 - 3 | 226 | 63 | ||
BC016584.1 | - | 2 - 3 | 239 | 3157800 | 63 | |
BC053729.1 | - | 2 - 3 | 182 | 30065192 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |