Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038604.1 | NP_001033693.1 | 4 - 5 | 394 | 99 | |
NM_021364.2 | NP_067339.1 | 3 - 4 | 319 | 99 | ||
GenBank mRNA | AF139769.1 | - | 3 - 4 | 289 | 99 | |
AK036697.1 | - | 3 - 4 | 285 | 99 | ||
AK046600.1 | - | 3 - 4 | 306 | 99 | ||
AK137482.1 | - | 3 - 4 | 320 | 99 | ||
BC104364.1 | - | 4 - 5 | 353 | 40049817 | 99 | |
BC104365.1 | - | 4 - 5 | 353 | 40049818 | 99 | |
CN670295.1 | - | 4 - 5 | 356 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |