Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200043402.mm.1 |
NM_001205076.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043402.mm.1 |
NM_001205076.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043402.mm.1 |
NM_021566.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043402.mm.1 |
NM_021566.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043402.mm.1 |
TC0200005089.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200043404.mm.1 |
NM_021566.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043404.mm.1 |
NM_021566.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043404.mm.1 |
TC0200005089.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200043403.mm.1 |
NM_001205076.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043403.mm.1 |
NM_001205076.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043403.mm.1 |
NM_021566.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043403.mm.1 |
NM_021566.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200043403.mm.1 |
TC0200005089.mm.1 |
MTA TC |