Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmfg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039192.1 | NP_001034281.1 | 3 - 4 | 255 | 70 | |
NM_001290550.1 | NP_001277479.1 | 2 - 3 | 181 | 70 | ||
NM_022024.2 | NP_071307.1 | 3 - 4 | 278 | 70 | ||
GenBank mRNA | AB003588.1 | - | 3 - 4 | 212 | 70 | |
AF297221.1 | - | 3 - 4 | 275 | 70 | ||
AK002834.1 | - | 3 - 4 | 279 | 70 | ||
AK009967.1 | - | 3 - 4 | 255 | 70 | ||
AK014214.1 | - | 3 - 4 | 279 | 70 | ||
BC011488.1 | - | 3 - 4 | 231 | 3581858 | 70 | |
BE629139.1 | - | 2 - 3 | 141 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |