Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpys | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164466.1 | NP_001157938.1 | 1 - 2 | 372 | 99 | |
NM_022722.3 | NP_073559.3 | 1 - 2 | 372 | 99 | ||
GenBank mRNA | AF227731.1 | - | 1 - 2 | 290 | 99 | |
AF249296.1 | - | 1 - 2 | 359 | 99 | ||
AK004822.1 | - | 1 - 2 | 372 | 99 | ||
AK004899.2 | - | 1 - 2 | 369 | 99 | ||
BC037086.1 | - | 1 - 2 | 341 | 5123860 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |