Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ciao1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025296.4 | NP_079572.2 | 2 - 3 | 477 | 74 | |
GenBank mRNA | AK002314.1 | - | 2 - 3 | 422 | 74 | |
AK028376.1 | - | 2 - 3 | 404 | 74 | ||
BC004089.1 | - | 2 - 3 | 398 | 3593736 | 74 | |
BI734652.1 | - | 2 - 3 | 477 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |