Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifrd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025903.2 | NP_080179.1 | 3 - 4 | 436 | 58 | |
GenBank mRNA | AK007687.1 | - | 3 - 4 | 437 | 58 | |
AK149496.1 | - | 3 - 4 | 417 | 58 | ||
AK156625.1 | - | 3 - 4 | 430 | 58 | ||
AK165967.1 | - | 3 - 4 | 429 | 58 | ||
AK166130.1 | - | 3 - 4 | 504 | 58 | ||
AK166258.1 | - | 3 - 4 | 394 | 58 | ||
AY298860.1 | - | 3 - 4 | 265 | 58 | ||
BC023486.1 | - | 3 - 4 | 384 | 58 | ||
BC037434.1 | - | 3 - 4 | 392 | 2651172 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |