Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27873630 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27873632 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27873631 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2084741 |
BC096688.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2084741 |
BC043075.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2084741 |
NM_026107.2 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
661588 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
329081 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
396949 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
650894 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
867102 |
AK014939.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
867102 |
AK030974.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
867102 |
AK034498.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
867102 |
AK040574.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
867102 |
AK047093.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
867102 |
AK147743.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
867102 |
BC031923.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
867102 |
NM_027590.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
867102 |
XM_977857.2 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
831750 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
538413 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
417495 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
423824 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
855131 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
579354 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
483461 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
39621 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P122237 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P139412 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2084741 |
BC096688.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2084741 |
BC043075.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2084741 |
NM_026107.2 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P337755 |
BC096688.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P337755 |
AK007308.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P337755 |
BC043075.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P337755 |
NM_026107.2 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27873599 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27873638 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27873622 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27873610 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27873628 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27873635 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27873601 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27873626 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27873641 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27873627 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27873623 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27873632 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27873600 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27873637 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27873595 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27873608 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27873614 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27873607 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27873636 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27873629 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27873613 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27873594 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27873619 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27873604 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27873616 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27873597 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27873618 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27873621 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27873596 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P122237 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P139412 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07375932 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07375904 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P337755 |
BC096688.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P337755 |
AK007308.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P337755 |
BC043075.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P337755 |
NM_026107.2 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P127048 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P139412 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P122237 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09096974 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09024310 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09024312 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09096977 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09024302 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09024309 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27873630 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27873632 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27873631 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873627 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873715 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873666 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873707 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873658 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873741 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873597 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873710 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873730 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873622 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873623 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873630 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873651 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873714 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873682 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873679 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873583 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873635 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873653 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873671 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873734 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873613 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873718 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873750 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873722 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873614 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873596 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873761 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873650 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873640 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873675 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873731 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873659 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873626 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873595 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873664 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873619 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873607 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873693 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873724 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873758 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873616 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873708 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873590 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873644 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873697 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873711 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873763 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873740 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873676 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873629 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873756 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873662 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873747 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873620 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873677 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873657 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873600 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873705 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873604 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873637 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873654 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873753 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873704 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873729 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873703 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873736 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873695 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873725 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873681 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873587 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873594 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873680 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873700 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873749 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873618 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873632 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873672 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873661 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873684 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873694 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27873610 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873629 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873610 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09339402 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873658 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09480978 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09252728 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873614 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09475071 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873618 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873657 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873594 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873601 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873661 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09320151 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873666 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09523806 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09385572 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873664 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873695 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09523804 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873596 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873613 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873602 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873707 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873641 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09290541 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873659 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09189261 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873622 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873636 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873627 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873647 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873623 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873758 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873642 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873632 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873626 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09523812 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873654 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873592 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873616 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873703 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873640 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09212138 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873608 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873621 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873704 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873600 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873638 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09240201 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873637 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873604 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873763 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873764 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873651 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09252732 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873643 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873630 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873646 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873662 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873697 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09517035 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873653 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09402100 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27873644 |
- |
- |