Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fahd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029629.2 | NP_083905.1 | 6 - 7 | 952 | 97 | |
GenBank mRNA | AI841210.1 | - | 2 - 3 | 97 | ||
AK002291.1 | - | 6 - 7 | 930 | 97 | ||
AK005134.1 | - | 5 - 6 | 1189 | 97 | ||
AK080868.1 | - | 6 - 7 | 938 | 97 | ||
AK170875.1 | - | 6 - 7 | 926 | 97 | ||
BC030182.1 | - | 6 - 7 | 903 | 5044017 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |