Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk3-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_004853.1 | 3 - 4 | 315 | 96 | ||
GenBank mRNA | AK009918.1 | - | 1 - 2 | 141 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |