Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10382785 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10382784 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10382780 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10382782 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10382786 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10382783 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10382787 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10382781 |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1460472_PM_at |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10382779 |
- |
- |
Mouse Genome 430 2.0 Array |
1460472_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1460472_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
69681_PM_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
69681_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR1100018302.mm.1 |
NR_004853.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018302.mm.1 |
TC1100001925.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100018304.mm.1 |
NR_004853.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100018304.mm.1 |
TC1100001925.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1460472_at |
- |
- |
Mouse Expression Set 430 |
1460472_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1460472_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1460472_PM_at |
- |
- |