Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhpn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027897.4 | NP_082173.3 | 10 - 11 | 1327 | 61 | |
GenBank mRNA | AK004849.1 | - | 10 - 11 | 1250 | 61 | |
AK050214.1 | - | 10 - 11 | 1241 | 61 | ||
BC100314.1 | - | 10 - 11 | 1234 | 30848276 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |