Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289543.1 | NP_001276472.1 | 5 - 6 | 901 | 58 | |
NM_001289544.1 | NP_001276473.1 | 6 - 7 | 1225 | 58 | ||
NM_028227.3 | NP_082503.2 | 6 - 7 | 1097 | 58 | ||
GenBank mRNA | AF321920.1 | - | 6 - 7 | 989 | 58 | |
AF321921.1 | - | 6 - 7 | 1226 | 58 | ||
AK013885.1 | - | 4 - 5 | 590 | 58 | ||
AK034212.1 | - | 1 - 2 | 30 | 58 | ||
AK143718.1 | - | 5 - 6 | 891 | 58 | ||
BC038490.1 | - | 3 - 4 | 466 | 3599718 | 58 | |
BC062932.1 | - | 6 - 7 | 1073 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |