Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029402.3 | NP_083678.2 | 6 - 7 | 704 | 69 | |
GenBank mRNA | AK004875.2 | - | 6 - 7 | 660 | 69 | |
AK016520.1 | - | 6 - 7 | 674 | 69 | ||
AK144693.1 | - | 6 - 7 | 677 | 69 | ||
AK160597.1 | - | 6 - 7 | 689 | 69 | ||
AK220287.1 | - | 6 - 7 | 679 | 69 | ||
BC026779.1 | - | 6 - 7 | 605 | 4018298 | 69 | |
BC027428.1 | - | 6 - 7 | 667 | 3983461 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |